Ethidium bromide
Glass bottle for solids, 1 g
RESERES3011E-A101X
: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
- :1239-45-8
- :C₂₁H₂₀BrN₃
- :MFCD00011724
- :394.31 g/mol
- :13,04751
- :72 °C (0.1 mmHg)
- :261…264 °C
- :1.01 g/cm³ (20 °C)
- :<100 °C
- :Ambient
- :214-984-6
- :2811
- :6.1,I
- Pack type:Glass bottle for solids
- Pk:1 g