Order Entry
Norway
Orders LinkContactUsLinkComponent
 

Test fluids for viscometers

S100* (silicone oil), ~100, 500 mL
Catalog # HAAK082-5373
Test fluids for viscometers
Catalog # HAAK082-5373
Supplier:  
CAS Number:  
Test fluids for viscometers

S100* (silicone oil), ~100, 500 mL

CAS Number:  
Discontinued
Product Details & Documents

Test and certificated fluids for viscometery - mesurement temperature at 20 °C unless indicated.

 : * Measurement temperature 25 °C

Specifications

Specifications

  • Pk:
    500 mL
  • Type:
    S100* (silicone oil)
  • Viscosity:
    ~100

Specifications