Order Entry
Norway
ContactUsLinkComponent
THERMOSTAT PROCESS INTEGRAL XT 1850 W | -50 ... 200C | HEATING CAPACITY: 10,4 KW | COOLING CAPACITY: 18,5 KW 1 * 1 items
Catalog # 461-2947
Supplier:  LAUDA
undefined
THERMOSTAT PROCESS INTEGRAL XT 1850 W | -50 ... 200C | HEATING CAPACITY: 10,4 KW | COOLING CAPACITY: 18,5 KW 1 * 1 items
Catalog # 461-2947
Supplier:  LAUDA