Order Entry
Norway
Orders LinkContactUsLinkComponent
Soil samplers, Mole

Soil samplers, Mole

Mole with handle and bar, 750 mm
Catalog # BURK5350-1005
Supplier:   BURKLE
Soil samplers, Mole
Catalog # BURK5350-1005
Supplier:  BURKLE
Soil samplers, Mole
Soil samplers, Mole

Mole with handle and bar, 750 mm

Price
kr 5 400,00
Log in for å se tilgjengelighet og deres pris
1 * 1 Items
Product Details & Documents

Very robust model with specially hardened forged drill bits. For determining soil properties, moisture and structure. The shape of the Mole‘s drill bits ensures minimum friction when drilling and removing the Mole from the ground.The Mole has different components. Handle with 75 cm rod, extension rod 100 cm if necessary and one of seven available drill bits, each of which is designed for a particular soil type. The components can be combined without any problems. Extension rods can extend the working depth of the Mole down to 5 m. Please order drill bits seperately.

  • Physical efforts are reduced to a minimum
Spesifikasjoner
  • Description:
    Mole with handle and bar
  • Length:
    750 mm
Accessories

Produktinformasjon

  • Catalog #
    Availability
    Price
  • Extension rod (100 cm)
    Extension rod (100 cm)
    BURK5350-1010
    Log in for å se tilgjengelighet og deres pris
    1 * 1 Items - kr 3 050,00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.