Methyl orange
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 547-58-0 Molecular Formula: C14H14N3NaO3S Formula Weight: 327.34 Odour: Weak, characteristic pH: 6.5 Melting Point: 300° Beilstein Reference: 4732884 Merck Reference: 14,6105 UN No.: 3143 MDL No.: MFCD00007502
Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine
- CAS Number:547-58-0
- Formula:C₁₄H₁₄N₃NaO₃S
- MDL Number:MFCD00007502
- Molecular Weight:327.34 g/mol
- Merck Index:14,6105
- Boiling Pt:∼90 °C (1013 hPa)
- Melting Pt:>300 °C
- Density:∼1 g/cm³ (20 °C)
- Flash Pt:38 °C
- Storage Temperature:Ambient
- EINECS:208-925-3
- UN:3143
- ADR:6.1,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



