Order Entry
Norway
Orders LinkContactUsLinkComponent
(±)-Linalool ≥97%

(±)-Linalool ≥97%

Glass bottle, 500 mL
Catalog # A14424.AP
Irritant
(±)-Linalool ≥97%
Catalog # A14424.AP
CAS Number:  78-70-6
(±)-Linalool ≥97%
(±)-Linalool ≥97%

Glass bottle, 500 mL

CAS Number:  78-70-6
Price
kr 1 825,00
Log in for availability
1 * 500 mL
Product Details & Documents

Purity: 97% CAS No.: 78-70-6 Molecular Formula: C10H18O Formula Weight: 154.25 Flash Point: 76°(168°F) Physical Form: Liquid Density: 0.865 Refractive Index: 1.4620 Appearance: Colourless Boiling Point: 199-200° Beilstein Reference: 1721488 Merck Reference: 14,5495 MDL No.: MFCD00008906

Synonyms:  (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
  • CAS Number:78-70-6
  • Formula:C₁₀H₁₈O
  • MDL Number:MFCD00008906
  • Molecular Weight:154.25 g/mol
  • Merck Index:14,5495
  • Boiling Pt:199…200 °C
  • Density:0.865
  • Flash Pt:76 °C (168 °F)
  • Storage Temperature:Ambient
  • EINECS:201-134-4
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.