Order Entry
Norway
Orders LinkContactUsLinkComponent
(±)-Linalool ≥97%

(±)-Linalool ≥97%

Glass bottle, 100 mL
Catalog # A14424.AE
Irritant
(±)-Linalool ≥97%
Catalog # A14424.AE
CAS Number:  78-70-6
(±)-Linalool ≥97%
(±)-Linalool ≥97%

Glass bottle, 100 mL

CAS Number:  78-70-6
Price
kr 500,00
Log in for availability
1 * 100 mL
Product Details & Documents

Purity: 97% CAS No.: 78-70-6 Molecular Formula: C10H18O Formula Weight: 154.25 Flash Point: 76°(168°F) Physical Form: Liquid Density: 0.865 Refractive Index: 1.4620 Appearance: Colourless Boiling Point: 199-200° Beilstein Reference: 1721488 Merck Reference: 14,5495 MDL No.: MFCD00008906

Synonyms:  (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
  • CAS Number:78-70-6
  • Formula:C₁₀H₁₈O
  • MDL Number:MFCD00008906
  • Molecular Weight:154.25 g/mol
  • Merck Index:14,5495
  • Boiling Pt:199…200 °C
  • Density:0.865
  • Flash Pt:76 °C (168 °F)
  • Storage Temperature:Ambient
  • EINECS:201-134-4
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications