o-Xylene ≥99%
Glass bottle, 100 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 95-47-6 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 32°(89°F) Refractive Index: 1.5050 Melting Point: -25° Boiling Point: 143-145° Beilstein Reference: 1815558 Merck Reference: 14,10081 UN No.: 1307 MDL No.: MFCD00008519
Synonyms: 1,2-Dimethylbenzene
- CAS Number:95-47-6
- Formula:C₈H₁₀
- MDL Number:MFCD00008519
- Molecular Weight:106.17 g/mol
- Merck Index:14,10081
- Boiling Pt:143…145 °C
- Melting Pt:–25 °C
- Density:0.877
- Flash Pt:32 °C (89 °F)
- Storage Temperature:Ambient
- EINECS:202-422-2
- UN:1307
- ADR:3,III
Documents
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Organic Photovoltaics
Product Literature
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL



