Order Entry
Norway
Orders LinkContactUsLinkComponent
o-Xylene ≥99%
Glass bottle, 2.500 mL
  A11358.0F
o-Xylene ≥99%
  A11358.0F
 :  95-47-6
o-Xylene ≥99%
o-Xylene ≥99%
Glass bottle, 2.500 mL
 :  95-47-6
kr 1 325,00
1 * 2.500 mL
Purity: 99% CAS No.: 95-47-6 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 32°(89°F) Refractive Index: 1.5050 Melting Point: -25° Boiling Point: 143-145° Beilstein Reference: 1815558 Merck Reference: 14,10081 UN No.: 1307 MDL No.: MFCD00008519
 :  1,2-Dimethylbenzene
  •  :95-47-6
  •  :C₈H₁₀
  •  :MFCD00008519
  •  :106.17 g/mol
  •  :14,10081
  •  :143…145 °C
  •  :–25 °C
  •  :0.877
  •  :32 °C (89 °F)
  •  :Ambient
  •  :202-422-2
  •  :1307
  •  :3,III
  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL