Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TARGET DP STORAGE CAP PTFE LINED 1 * 100 items
Catalog # NSCAC4000-99
Supplier:
THERMO NATIONAL SCIENTIFIC
Expand image
TARGET DP STORAGE CAP PTFE LINED 1 * 100 items
Catalog # NSCAC4000-99
Supplier:
THERMO NATIONAL SCIENTIFIC