Order Entry
Norway
Orders LinkContactUsLinkComponent
Microtube racks

Microtube racks

24 (2×12), 0,5 ml, 265×50×50 mm
Catalog # 211-8065
Microtube racks
Catalog # 211-8065
Microtube racks
Microtube racks

24 (2×12), 0,5 ml, 265×50×50 mm

Price
kr 910,00
Log in for å se tilgjengelighet og deres pris
1 * 1 Items
Product Details & Documents

Acrylic.

Spesifikasjoner
  • For tubes:
    0,5 ml
  • No. of Holes:
    24 (2×12)
  • Seq:
    10
  • W×D×H:
    265×50×50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.