Order Entry
Norway
Orders LinkContactUsLinkComponent
Microtube racks

Microtube racks

48 (4×12), 0,5 ml, 265×100×50 mm
Catalog # 211-8066
Microtube racks
Catalog # 211-8066
Microtube racks
Microtube racks

48 (4×12), 0,5 ml, 265×100×50 mm

Price
kr 1 825,00
Log in for availability
1 * 1 Items
Product Details & Documents

Acrylic.

Specifications
  • For tubes:
    0,5 ml
  • No. of Holes:
    48 (4×12)
  • Seq:
    11
  • W×D×H:
    265×100×50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.