Microtube racks
24 (2×12), 1,5 ml, 265×50×50 mm
Catalog # 211-8067
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Gå til:
Product Details & Documents
Acrylic.
Spesifikasjoner
- For tubes:1,5 ml
- No. of Holes:24 (2×12)
- Seq:13
- W×D×H:265×50×50 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



