Order Entry
Norway
Orders LinkContactUsLinkComponent
Microtube racks

Microtube racks

12 (2×6), 1,5 ml, 135×50×50 mm
Catalog # 211-8068
Microtube racks
Catalog # 211-8068
Microtube racks
Microtube racks

12 (2×6), 1,5 ml, 135×50×50 mm

Price
kr 720,00
Log in for å se tilgjengelighet og deres pris
1 * 1 Items
Product Details & Documents

Acrylic.

Spesifikasjoner
  • For tubes:
    1,5 ml
  • No. of Holes:
    12 (2×6)
  • Seq:
    12
  • W×D×H:
    135×50×50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.