0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Thermoplastic quick coupling, Acetal, female, connecting 8 / 6 mm 1 * 1 items
Catalog # 201-7474
Supplier:
KOEHLER TECHNISCHE PRODUKTEN
Expand image
Thermoplastic quick coupling, Acetal, female, connecting 8 / 6 mm 1 * 1 items
Catalog # 201-7474
Supplier:
KOEHLER TECHNISCHE PRODUKTEN