Order Entry
Norway
Orders LinkContactUsLinkComponent
Chlorhexidine, Reference Standards

Chlorhexidine, Reference Standards

Chlorhexidine Acetate, USP reference standard, Vial, 200 mg
Catalog # 1111103.
Supplier:   USP
Chlorhexidine, Reference Standards
Catalog # 1111103.
Supplier:  USP
CAS Number:  
Chlorhexidine, Reference Standards

Chlorhexidine Acetate, USP reference standard, Vial, 200 mg

CAS Number:  
Price
kr 4 600,00
Log in for availability
1 * 200 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • Description:
    Chlorhexidine Acetate
  • UN No.:
    2811
  • Cat. No.:
    1111103.
  • Formula:
    C₂₆H₃₈Cl₂N₁₀O₄
  • Description:
    Chlorhexidine Acetate
  • CAS No.:
    56-95-1

Specifications