Order Entry
Norway
Orders LinkContactUsLinkComponent
Progesterone, Reference Standards
Progesterone, Reference Standards
Progesterone, USP reference standard, Vial, 200 mg
  1568007.
 :   USP
Progesterone, Reference Standards
  1568007.
 :  USP
 :  
Progesterone, Reference Standards
Progesterone, USP reference standard, Vial, 200 mg
 :  
kr 3 825,00
1 * 200 mg
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Progesterone System Suitability Mixture: Mixture contains 20-α-Progerol, 20-β-Progerol, 17α-Progesterone, 20-α-Progerol acetate, 20-β-Progerol acetate and Cyclohexylidene progesterone in Progesterone
Certifications: US Pharmacopea Reference Standard
  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • Description:
    Progesterone
  • Cat. No.:
    1568007.
  • Description:
    Progesterone
  • CAS No.:
    57-83-0