Order Entry
Norway
Orders LinkContactUsLinkComponent
3-(2-Hydroxyethyl)indole

3-(2-Hydroxyethyl)indole

Glass bottle for solids, 5 g
Catalog # 8.41379.0005
Supplier:   Merck
3-(2-Hydroxyethyl)indole
Catalog # 8.41379.0005
Supplier:  Merck
CAS Number:  526-55-6
3-(2-Hydroxyethyl)indole

Glass bottle for solids, 5 g

CAS Number:  526-55-6
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  3-Indoleethanol, Indol-3-ethanol, Tryptophol, β-Indol-3-ylethanol
  • CAS Number:526-55-6
  • Formula:C₁₀H₁₁NO
  • MDL Number:MFCD00005659
  • Molecular Weight:161.2 g/mol
  • Merck Index:12,09930
  • Boiling Pt:358 °C (1013 hPa)
  • Melting Pt:55…57 °C
  • EINECS:208-393-2

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Cat. No.:
    8.41379.0005

Specifications