Order Entry
Norway
Orders LinkContactUsLinkComponent
ADA

ADA

Glass bottle for solids, 100 g
Catalog # MOLE15676222-100G
Marketsource
Supplier:   Molekula
ADA
Catalog # MOLE15676222-100G
Supplier:  Molekula
Marketsource
CAS Number:  26239-55-4
ADA

Glass bottle for solids, 100 g

CAS Number:  26239-55-4
Price
kr 2 075,00
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  2,2'-[(2-Amino-2-oxoethyl)imino]diacetic acid, N-(2-Acetamido)iminodiacetic acid, 2,2'-((2-Amino-2-oxoethyl)azanediyl)diacetic acid
  • CAS Number:26239-55-4
  • Formula:H₂NC(O)CH₂N(CH₂COOH)₂
  • MDL Number:MFCD00008031
  • Molecular Weight:190.16 g/mol
  • Merck Index:13,00148
  • Melting Pt:217…218 °C
  • Storage Temperature:Ambient
  • EINECS:247-530-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications