Gå til Overskrift
Gå til Hovedinnhold
Gå til Bunntekst
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Refractive Index AAALiquid 1,335 1 * 1 Items
Catalog # CARG1803X1.335
Supplier:
CARGILLE
Expand image
Refractive Index AAALiquid 1,335 1 * 1 Items
Catalog # CARG1803X1.335
Supplier:
CARGILLE