kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Refractive Index Liquid Series B - 1/2 1 * 1 Items
CARG18122
:
CARGILLE
Refractive Index Liquid Series B - 1/2 1 * 1 Items
CARG18122
:
CARGILLE