Order Entry
Norway
Orders LinkContactUsLinkComponent
Coil spring
Coil spring
Spiral spring 30 N/m
Catalog # 765-1153
Marketsource
Supplier:   PHYWE
Coil spring
Catalog # 765-1153
Supplier:  PHYWE
Marketsource
Coil spring
Coil spring
Spiral spring 30 N/m
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
Coil springs made of spring steel. With stirrups at both ends. Suitable for Hook's team and swing attempts.
Specifications
  • Description:
    Spiral spring 30 N/m