Order Entry
Norway
Orders LinkContactUsLinkComponent
Coil spring
Coil spring
Spiral spring 3 N/m
Catalog # 765-1151
Marketsource
Supplier:   PHYWE
Coil spring
Catalog # 765-1151
Supplier:  PHYWE
Marketsource
Coil spring
Coil spring
Spiral spring 3 N/m
Price
kr 255,00
Log in for availability
1 * 1 Items
Product Details & Documents
Coil springs made of spring steel. With stirrups at both ends. Suitable for Hook's team and swing attempts.
Specifications
  • Description:
    Spiral spring 3 N/m