Tripod rods, square, PASS
10 mm, Tripod rod 450 mm, 400 mm
Catalog # 765-1141
Marketsource
Supplier: PHYWE
Jump to:
Product Details & Documents
Tripod rods with square cross section 12×12 mm. Made of galvanized galvanized steel pipe. One end of the rod is provided with a cylindrical rod (50×10 mm). The other end closed. Square rods provide unmatched torsional rigidity compared to cylindrical frame rods.
Specifications
- Ø:10 mm
- Description:Tripod rod 450 mm
- Length:400 mm



