Pendulum bobs
Steel pendulum bob, 25,4 mm
Catalog # 765-1155
Marketsource
Supplier: PHYWE
Jump to:
Product Details & Documents
Suitable for use as pendulum bobs for frequency experiments. Hardened and polished ball bearings with eyelet.
Specifications
- Description:Steel pendulum bob
- Ø:25,4 mm