Order Entry
Norway
Orders LinkContactUsLinkComponent
BOD Nutrient Buffer Pillows, 4 ml size for 4 liter Dilution Water, pk/50, used in HACH Method 8043 ans 10230, BOD determination usin g Dilution Method 1 * 50 items
BOD Nutrient Buffer Pillows, 4 ml size for 4 liter Dilution Water, pk/50, used in HACH Method 8043 ans 10230, BOD determination usin g Dilution Method 1 * 50 items
Catalog # HACH2436466
Supplier:   Hach
BOD Nutrient Buffer Pillows, 4 ml size for 4 liter Dilution Water, pk/50, used in HACH Method 8043 ans 10230, BOD determination usin g Dilution Method 1 * 50 items
Catalog # HACH2436466
Supplier:  Hach
BOD Nutrient Buffer Pillows, 4 ml size for 4 liter Dilution Water, pk/50, used in HACH Method 8043 ans 10230, BOD determination usin g Dilution Method 1 * 50 items
BOD Nutrient Buffer Pillows, 4 ml size for 4 liter Dilution Water, pk/50, used in HACH Method 8043 ans 10230, BOD determination usin g Dilution Method 1 * 50 items
Price
kr 1 125,00
Log in for availability
1 * 50 Items