Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Ordreinnføring
Ordrestatus
Din konto
Handlevogn
Norway
Ordreinnføring
Ordrestatus
Din konto
Handlevogn
ContactUsLinkComponent
Endotoxin Removal 1 * 1 items
Katalog # ROCKCUST47
Supplier:
Rockland Immunochemicals
Expand image
Endotoxin Removal 1 * 1 items
Katalog # ROCKCUST47
Supplier:
Rockland Immunochemicals