Order Entry
Norway
Orders LinkContactUsLinkComponent
Staining systems, EasyDip™

Staining systems, EasyDip™

Slide staining cuvettes, White, Acetal, 76×64×92 mm
Catalog # 720-0796
Supplier:   Simport Scientific
Staining systems, EasyDip™
Catalog # 720-0796
Supplier:  Simport Scientific
Staining systems, EasyDip™
Staining systems, EasyDip™

Slide staining cuvettes, White, Acetal, 76×64×92 mm

Price
kr 1 450,00
Log in for availability
1 * 6 Items
Product Details & Documents

The slide staining system is made of acetal plastic.

  • Jars can be loosely joined to each other to keep the same order when moved around
  • Wide base for stability while inside is recessed to reduce reagent volume
  • Available in five colours to aid identification of contents or applications

The slide staining system has two components: A square staining jar and a 12 position vertical slide rack. The staining jar is made of resistant acetal plastic and will resist most staining agents including alcohol and xylene but not phenol. Autoclavable, it is therefore suitable for special stains, frozen sections and special processes.

Volume: 80 ml

W×D×H: 76×64×92 mm

Ordering information: The staining jars and staining rack are sold separately, except for the assorted package (720-0791), which contains one jar of each colour (blue, green, pink, white, yellow) and a rack. The EasyDip aluminum holder is designed to hold six EasyDip slide staining jars. The EasyDip slide staining kit (SIMPM906-12AS) contains 6 jars (blue, green, pink, 2xwhite, yellow), an anodized aluminium rack, and one slide staining rack.

Specifications
  • Colour:
    White
  • Description:
    Slide staining cuvettes
  • Material:
    Acetal
  • Seq:
    05
  • W×D×H:
    76×64×92 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.