Order Entry
Norway
Orders LinkContactUsLinkComponent
Phloretin ≥98%

Phloretin ≥98%

Glass bottle for solids, 25 mg
Catalog # L10991.MB
Irritant
Phloretin ≥98%
Catalog # L10991.MB
CAS Number:  60-82-2
Phloretin ≥98%
Phloretin ≥98%

Glass bottle for solids, 25 mg

CAS Number:  60-82-2
Price
kr 630,00
Log in for availability
1 * 25 mg
Product Details & Documents

Purity: 98% CAS No.: 60-82-2 Molecular Formula: C15H14O5 Formula Weight: 274.28 Storage Temperature: 2°C to 8°C Melting Point: ca 263° dec. Beilstein Reference: 1887240 Merck Reference: 14,7326 MDL No.: MFCD00002288

  • CAS Number:60-82-2
  • Formula:C₁₅H₁₄O₅
  • MDL Number:MFCD00002288
  • Molecular Weight:274.28 g/mol
  • Merck Index:14,7326
  • Melting Pt:∼ 263 °C (dec.)
  • EINECS:200-488-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.