Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
96 WELL (U) NS NP W/O C 100/CS 1 * 1 Each
Catalog # ICNA97631105
Supplier:
MP Biomedicals
Expand image
96 WELL (U) NS NP W/O C 100/CS 1 * 1 Each
Catalog # ICNA97631105
Supplier:
MP Biomedicals