Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CINNARIZINE (1-TRANS-CINNAMYL-4-DIPHENYL 1 * 10 g
Catalog # ICNA0215498610
Supplier:
MP Biomedicals
Expand image
CINNARIZINE (1-TRANS-CINNAMYL-4-DIPHENYL 1 * 10 g
Catalog # ICNA0215498610
Supplier:
MP Biomedicals