Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TRP-NLE-ARG-PHE AMIDE 1 * 1 mg
Catalog # ICNA215318501
Supplier:
MP Biomedicals
Expand image
TRP-NLE-ARG-PHE AMIDE 1 * 1 mg
Catalog # ICNA215318501
Supplier:
MP Biomedicals