kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
D-(-)-ALPHA-PHENYLGLYCINE 1 * 25 g
ICNA215183225
:
MP Biomedicals
D-(-)-ALPHA-PHENYLGLYCINE 1 * 25 g
ICNA215183225
:
MP Biomedicals