kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
FURAN UN2389I (NOT FOR EXPORT) 1 * 100 g
ICNA215117580
:
MP Biomedicals
FURAN UN2389I (NOT FOR EXPORT) 1 * 100 g
ICNA215117580
:
MP Biomedicals