Single - component oxygenates solutions
Ethanol, with 10000 µg/ml in DI water, US EPA 502.2, 8021, 8206B, 1 ml/ampoule, 1
Catalog # REST30466
Supplier: Restek
Jump to:
Product Details & Documents
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specifications
Specifications
- Pk:1
- Standards:US EPA 502.2, 8021, 8206B
- Description:Ethanol, with 10000 µg/ml in DI water
- Pk Info:1 ml/ampoule
- Cat. No.:REST30466
- Formula:C₂H₅OH
- Description:Ethanol
- CAS No.:64-17-5
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



