Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SERUM,BOVINE CADET(10 X 100 MLS=$121.00) 1 * 100 ml
Catalog # ICNA219135180
Supplier:
MP Biomedicals
Expand image
SERUM,BOVINE CADET(10 X 100 MLS=$121.00) 1 * 100 ml
Catalog # ICNA219135180
Supplier:
MP Biomedicals