kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
ALPHA-BUNGAROTOXIN (125I)40 MCI/MG N 1 * 1 Each
ICNA6805201
:
MP Biomedicals
ALPHA-BUNGAROTOXIN (125I)40 MCI/MG N 1 * 1 Each
ICNA6805201
:
MP Biomedicals