Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Bp Liquid Paraffin Disc (TINT 296490 ) 1 * 1 items
TINT296490
:
Lovibond Tintometer
Bp Liquid Paraffin Disc (TINT 296490 ) 1 * 1 items
TINT296490
:
Lovibond Tintometer