Order Entry
Norway
Orders LinkContactUsLinkComponent
Test Strips, Zinc

Test Strips, Zinc

AQUADUR® 4-21, < 54 • >72 • >126 • >252 • >378 - ppm - CaCO₃; <3 • >4 • >7 • >14 • >21 - °e, 210×210×135 mm (packaging), 1000 Tests, 1.000
Catalog # MANA91224
Supplier:   MACHEREY-NAGEL
Test Strips, Zinc
Catalog # MANA91224
Supplier:  MACHEREY-NAGEL
CAS Number:  
Test Strips, Zinc
Test Strips, Zinc

AQUADUR® 4-21, < 54 • >72 • >126 • >252 • >378 - ppm - CaCO₃; <3 • >4 • >7 • >14 • >21 - °e, 210×210×135 mm (packaging), 1000 Tests, 1.000

CAS Number:  
Price
kr 5 140,00
Log in for å se tilgjengelighet og deres pris
1 * 1.000 Items
Product Details & Documents

QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. These test strips are immediately ready-to-use. They do not require additional accessories. The test strips are intended for single use, maintenance or calibration are not required.

  • Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

These test strips allow the quick and easy determination of Zinc in solutions. They come with all necessary reagents so that the measurement can be started immediately. A reliable result is obtained within 2 minutes.

Spesifikasjoner

Spesifikasjoner

  • Pk:
    1.000
  • Description:
    AQUADUR® 4-21
  • Dimensions:
    210×210×135 mm (packaging)
  • Tests per kit:
    1000 Tests
  • Range:
    < 54 • >72 • >126 • >252 • >378 - ppm - CaCO₃; <3 • >4 • >7 • >14 • >21 - °e
  • Cat. No.:
    MANA91224
  • Parameters:
    All reagents included
  • Contents:
    1000 individually sealed test strips in a box
  • Platform:
    Semi-quantitative test strips
  • Compliance:
    No hazardous material
  • Instrument:
    No
  • Shelf life (months):
    12 (from production)
  • Method:
    Total hardness - green → red
  • Weight:
    598,2 g

Spesifikasjoner

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.