Order Entry
Norway
Orders LinkContactUsLinkComponent
 

VISOCOLOR® Colorimetric Test, Nitrite

VISOCOLOR® ECO Nitrite, refill pack, test no.5-44; Measuring range on PF-12Plus. Range on other photometers can be different, Reagents for 120 tests in a box, 0 • 0,02 • 0,03 • 0,05 • 0,07 • 0,1 • 0,2 • 0,3 • 0,5 mg/L(NO₂⁻); 0,02 - 0,50 mg/L (NO₂⁻), Sulfanilic acid/1, PF-12Plus; PF-3 Fish; Advance; Visual, 170×115×70 mm (including packaging), Hazardous material: Yes, 120, 1 KIT
Catalog # MANA931244
Supplier:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Nitrite
Catalog # MANA931244
Supplier:  MACHEREY-NAGEL
CAS Number:  
VISOCOLOR® Colorimetric Test, Nitrite
VISOCOLOR® Colorimetric Test, Nitrite

VISOCOLOR® ECO Nitrite, refill pack, test no.5-44; Measuring range on PF-12Plus. Range on other photometers can be different, Reagents for 120 tests in a box, 0 • 0,02 • 0,03 • 0,05 • 0,07 • 0,1 • 0,2 • 0,3 • 0,5 mg/L(NO₂⁻); 0,02 - 0,50 mg/L (NO₂⁻), Sulfanilic acid/1, PF-12Plus; PF-3 Fish; Advance; Visual, 170×115×70 mm (including packaging), Hazardous material: Yes, 120, 1 KIT

CAS Number:  
Price
kr 566,00
Log in for availability
1 * 1 Each
Specifications

Specifications

  • Chemical method:
    Sulfanilic acid/1
  • Pk:
    1 KIT
  • Description:
    VISOCOLOR® ECO Nitrite, refill pack, test no.5-44; Measuring range on PF-12Plus. Range on other photometers can be different
  • Dimensions:
    170×115×70 mm (including packaging)
  • Type:
    Hazardous material: Yes
  • For unit:
    PF-12Plus; PF-3 Fish; Advance; Visual
  • Tests per kit:
    120
  • Range:
    0 • 0,02 • 0,03 • 0,05 • 0,07 • 0,1 • 0,2 • 0,3 • 0,5 mg/L(NO₂⁻); 0,02 - 0,50 mg/L (NO₂⁻)
  • Contents:
    Reagents for 120 tests in a box
  • Cat. No.:
    MANA931244
  • Platform:
    Colorimetric test kits
  • Model:
    MANA931244
  • Shelf life (months):
    18
  • Storage temperature:
    15…25 °C
  • Weight:
    113,8 g (including packaging)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.