Order Entry
Norway
Orders LinkContactUsLinkComponent
 

VISOCOLOR® Colorimetric Tests, DEHA

VISOCOLOR® ECO DEHA; Carbohydrazide (carbo), hydroquinone (hydro), isoascorbic acid (ISA) and methylethyl ketone oxime (MEKO) are only photometrically evaluable, Reagents for 125 tests, colour chart and accessories in a box, 0 • 0,01 • 0,03 • 0,05 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 mg/L (DEHA); 0,005 - 0,5 mg/L (DEHA); 0,01 - 0,60 mg/L (Carbo); 0,01 - 0,60 mg/L (Carbo); 0,01 - 0,60 mg/L (Carbo); 0,01 - 1,00 mg/L (Hydro); 0,01 - 1,00 mg/L (Hydro); 0,025 - 1,500 mg/L (ISA), Redox reaction, PF-12Plus; Advance; Visual, 170×125×48 mm (Including packaging), 125, 1 KIT
Catalog # MANA931024
Supplier:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Tests, DEHA
Catalog # MANA931024
Supplier:  MACHEREY-NAGEL
CAS Number:  
VISOCOLOR® Colorimetric Tests, DEHA
VISOCOLOR® Colorimetric Tests, DEHA

VISOCOLOR® ECO DEHA; Carbohydrazide (carbo), hydroquinone (hydro), isoascorbic acid (ISA) and methylethyl ketone oxime (MEKO) are only photometrically evaluable, Reagents for 125 tests, colour chart and accessories in a box, 0 • 0,01 • 0,03 • 0,05 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 mg/L (DEHA); 0,005 - 0,5 mg/L (DEHA); 0,01 - 0,60 mg/L (Carbo); 0,01 - 0,60 mg/L (Carbo); 0,01 - 0,60 mg/L (Carbo); 0,01 - 1,00 mg/L (Hydro); 0,01 - 1,00 mg/L (Hydro); 0,025 - 1,500 mg/L (ISA), Redox reaction, PF-12Plus; Advance; Visual, 170×125×48 mm (Including packaging), 125, 1 KIT

CAS Number:  
Price
kr 2 830,00
Log in for availability
1 * 1 Each
Product Details & Documents

Reagents for the determination of DEHA in water samples.

  • Combines simple implementation with high measurement reliability

Simple determination directly on site or in the laboratory.

Sea water analysis: Yes

Hazardous material: No

Specifications

Specifications

  • Chemical method:
    Redox reaction
  • Pk:
    1 KIT
  • Description:
    VISOCOLOR® ECO DEHA; Carbohydrazide (carbo), hydroquinone (hydro), isoascorbic acid (ISA) and methylethyl ketone oxime (MEKO) are only photometrically evaluable
  • Dimensions:
    170×125×48 mm (Including packaging)
  • For unit:
    PF-12Plus; Advance; Visual
  • Tests per kit:
    125
  • Range:
    0 • 0,01 • 0,03 • 0,05 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 mg/L (DEHA); 0,005 - 0,5 mg/L (DEHA); 0,01 - 0,60 mg/L (Carbo); 0,01 - 0,60 mg/L (Carbo); 0,01 - 0,60 mg/L (Carbo); 0,01 - 1,00 mg/L (Hydro); 0,01 - 1,00 mg/L (Hydro); 0,025 - 1,500 mg/L (ISA)
  • Contents:
    Reagents for 125 tests, colour chart and accessories in a box
  • Cat. No.:
    MANA931024
  • Platform:
    Colorimetric test kits
  • Model:
    MANA931024
  • Shelf life (months):
    12
  • Storage temperature:
    15...25 °C
  • Weight:
    221 g (Including packaging)

Specifications