Order Entry
Norway
Orders LinkContactUsLinkComponent
 

Bismuth(III) oxide, Supelco®

Plastic bottle, 1 kg
Catalog # 1.01862.1000
Irritant
Supplier:   Merck
Bismuth(III) oxide, Supelco®
Catalog # 1.01862.1000
Supplier:  Merck
CAS Number:  1304-76-3
Bismuth(III) oxide, Supelco®

Plastic bottle, 1 kg

CAS Number:  1304-76-3
Price
kr 3 820,00
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Bismuth Trioxide, Dibismuth trioxide
  • CAS Number:1304-76-3
  • Formula:BiO₃
  • MDL Number:MFCD00003462
  • Molecular Weight:465.96 g/mol
  • Merck Index:13,01273
  • Boiling Pt:1890 °C (1013 hPa)
  • Melting Pt:825 °C
  • Density:8.9 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:215-134-7
Specifications

Specification Test Results

  • Melting Point:
    820 °C
  • Molar Mass:
    465.95 g/mol
  • Bulk Density:
    1000 kg/m3
  • Boiling Point:
    1890 °C
  • Density:
    8.93 g/cm3 (20 °C)

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg

Specifications