Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STORAGE SET 32.0 CU.FT. FREESER UPRIGHT 1 * 1 items
Catalog # REVC7189-1
Supplier:
REVCO TECHNOLOGIES
Expand image
STORAGE SET 32.0 CU.FT. FREESER UPRIGHT 1 * 1 items
Catalog # REVC7189-1
Supplier:
REVCO TECHNOLOGIES