TMB SUBSTRATE SOLUTION 1 * 100 mL
PIERN301
Frequently Bought Together
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
MicroWell™ Plates, 96 well, Nunc-Immuno™
: Thermo Fisher Scientific
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
: VWR Chemicals
Serological pipettes, Stripette™
: Corning
Ethanol, GPR RECTAPUR®
: VWR Chemicals