Order Entry
Norway
Orders LinkContactUsLinkComponent
Trays
350×250×70 mm
  BOCH8736
 :   BOCHEM
Trays
  BOCH8736
 :  BOCHEM
Trays
Trays
350×250×70 mm
kr 1 475,00
1 * 1 Items
Tray made of stainless steel with two handles.
  • Very good temperature resistance
  • Very good chemical resistance
  • Easy to clean
Multipurpose trays for a wide range of uses.
  • Seq:
    02
  • W×D×H:
    350×250×70 mm