Order Entry
Norway
Orders LinkContactUsLinkComponent
 

Overhead stirrer tools, paddle stirrers

70 mm, 8 mm, 400 mm
  BOCH9404
 :   BOCHEM
Overhead stirrer tools, paddle stirrers
  BOCH9404
 :  BOCHEM
Overhead stirrer tools, paddle stirrers
Overhead stirrer tools, paddle stirrers

70 mm, 8 mm, 400 mm

 
kr 840,00
   
1 * 1 Items
 

Paddle stirrers made of stainless steel, with three holes.

  • Ideal for stirring liquids at middle speed range
  • Suitable for stirring low to medium viscous media
  • Creates radial flow
 
  • Seq:
    02
  • Shaft Ø:
    8 mm
  • Shaft length:
    400 mm
  • Stirrer Ø:
    70 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.