0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TUBE SWT4 254NM, 4 WATT 1 * 1 items
Catalog # 730-1267
Supplier:
Syngene (a division of the Synoptics group)
Utvid bildet
TUBE SWT4 254NM, 4 WATT 1 * 1 items
Catalog # 730-1267
Supplier:
Syngene (a division of the Synoptics group)