0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
DENSIMET W THERM 1,00-1,02 0,0002 20C 1 * 1 items
Catalog # GERI0193S
Supplier:
GERING
Expand image
DENSIMET W THERM 1,00-1,02 0,0002 20C 1 * 1 items
Catalog # GERI0193S
Supplier:
GERING