Microscope slides
Objektglass, skårne, uten mattfelt, sodalimeglass, Hvit, 1 mm, 76×26 mm
Supplier: MARIENFELD
Jump to:
Product Details & Documents
Slides designed for in vitro diagnostic (IVD) applications.
- Pre-washed and ready to use
- Either ground or cut versions
- Soda-lime glass, Hydrolytic Class 3
Extra-white slides are used for low auto-fluorescence applications.
Documents
Specifications
- Farge:Hvit
- Beskrivelse:Objektglass, skårne, uten mattfelt, sodalimeglass
- L×B:76×26 mm
- Seq:01
- Tykkelse:1 mm



