0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Sil 180 Silicone Oil Bath Liquid, Temperature Range -40c to 200C, 5L 1 * 5 l
Catalog # 461-0020
Supplier:
HAAKE
Expand image
Sil 180 Silicone Oil Bath Liquid, Temperature Range -40c to 200C, 5L 1 * 5 l
Catalog # 461-0020
Supplier:
HAAKE